missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Dibenzyl Ester Silver Salt, TRC

Product Code. 30428203
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428203 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428203 Supplier Toronto Research Chemicals Supplier No. TRCP3594052.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphoric Acid Dibenzyl Ester Silver Salt
CAS 50651-75-7
Molecular Formula C14H14AgO4P
Quantity 2.5 g
Synonym Dibenzyl Hydrogen Phosphate Silver Salt, Dibenzyl Phosphate Silver Salt, Dibenzylphosphoric Acid Silver Salt
InChI Formula InChI=1S/C14H15O4P.Ag/c15-19(16,17-11-13-7-3-1-4-8-13)18-12-14-9-5-2-6-10-14;/h1-10H,11-12H2,(H,15,16);/q;+1/p-1
SMILES [Ag+].[O-]P(=O)(OCc1ccccc1)OCc2ccccc2
IUPAC Name silver;dibenzyl phosphate
Molecular Weight (g/mol) 385.1
Formula Weight 383.9681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.