missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phosphothreonine Monoclonal Antibody (PT-5H5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ 139200
This item is not returnable.
View return policy
Description
Phosphothreonine Monoclonal Antibody for Western Blot, ELISA
Phosphothreonine is a phosphoamino acid, the phosphorylated ester of threonine. There are three amino acids that are typically phosphorylated in eukaryotes: serine, threonine, and tyrosine. Threonine residues in endogenous proteins undergo phosphorylation through the action of a threonine kinase. Small amounts of free phosphothreonine can be detected in urine Antibodies against phosphothreonine can be used as an identifier in the analysis of peptides, proteins, and enzymes. [National Center for Biotechnology Information.
Specifications
| Phosphothreonine | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Mouse | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| PT-5H5 | |
| Unconjugated | |
| C4H10NO6P; O-phospho-L-threonine; Phospho-threonine; pThr; Threonine | |
| Phosphothreonine containing proteins. | |
| 100 μg | |
| Primary | |
| -20°C | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction