missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phosphotyrosine Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143742
This item is not returnable.
View return policy
Description
This antibody detects phosphotyrosine-containing proteins on SDS-PAGE immunoblots of EGF treated A431 cells, as well as pervanadate treated A431, HeLa, Jurkat, and endothelial cells.
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
Specifications
| Phosphotyrosine | |
| Polyclonal | |
| Unconjugated | |
| Pdpk1 | |
| Phosphotyrosine; pTyr; pY | |
| Anti-Phosphotyrosine was generated from a panel of phosphotyrosine-containing peptide immunogens designed from human protein sequences. All peptide sequences used are highly conserved phosphotyrosine motifs. | |
| 100 μL | |
| Primary | |
| Chemical, Multi-species, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| O15530, O55173, Q9Z2A0 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18607, 5170, 81745 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction