Learn More
WTW™ pHotoFlex™ Turb Set
Description
- Includes an infrared light source for nephelometric turbidity measurement
- Acc. to DIN 27027/ISO 7027
- Precision is comparable to laboratory instruments for turbidity measurement
- Together with the AMCO Clear™ standards, highest precision for the sensitive testing of drinking water is given
- Integrated pH measurement
- Automatic temperature compensation
- Field case set makes for an in-field laboratory with an integrated tray to hold the photometer, cuvettes, measuring beaker and a stand for the pH electrode SenTix™ 41
- LSdata for convenient data management and definition of user-defined methods
- 100 user-defined methods
- Data storage for 1000 data sets
- 0 to 16 pH range
- 0 to 1100 NTU/FNU turbidity range
- RS 232 interface, USB via optional adapter
Specifications
Specifications
| Product Type | CombiCheck COD Chemical oxygen demand O2 cell test kit |
| Accuracy | < 2nm wavelength accuracy, 0.005 abs. reproducibility; ±0.01 pH; 0.01 NTU/FNU or +2% of the measured value |
| For Use With (Application) | Water Analysis and Routine Measurement, pH and Turbidity Measurement |
| Includes | Field case with tray to hold instrument, calibration standard kit, LSdata and accessories |
| Light Source | LED |
| Wavelength | 436, 517, 557, 594, 610, 690, + 860 |