Learn More
Brady™ Photoluminescent Self-adhesive Dots
Codice prodotto. 11761316
Cambia vista
Click to view available options
Diameter (Metric):
16 mm
25 mm
Dimensione della confezione:
Pack of 15
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diameter (Metric) unitSize
11761316 16 mm Pack of 40
11766915 25 mm Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11761316 Fornitore Brady™ N. del fornitore 263382
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Use these Photoluminescent Self-adhesive Dots to mark anything that is important to locate, especially in complete darkness.
  • Use these photoluminescent dots to mark anything that is important to locate, especially in complete darkness.
Specifica
Description Photoluminescent individual dots (Self-adhesive)
Adhesive Type Self-adhesive
Coating None
Color Photoluminescent
Display Blank
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology None
Product Type Photoluminescent Tape
Sterility Non-sterile
No. per Pack 40 Pieces/Card
Quantity 40 Pk.
Shape Round
Dimensions (L x W) 16 x 16 mm
Length (English) 1.6 in.
Width (English) 1.6 in.
Width (Metric) 16.00 mm
Diameter (English) 0.63 in.
Diameter (Metric) 16 mm
Weight (Metric) 0.01 kg
Alternate Part Number PHOTOLUM DISCS-DIA 1,6
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema