Learn More
WTW™ Photometry Cuvette
Product Code. 11701333
Change view
Click to view available options
Volume (Metric):
3 mL
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11701333 3 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11701333 Supplier WTW™ Supplier No. 250621
Only null left
Add to basket
Add to basket
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
TRUSTED_SUSTAINABILITY
Specifikationer
Volume (Metric) 3 mL
For Use With (Equipment) Photometry
Storage Requirements 15°C to 25°C
Type TOC Cell Test Kit
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.