missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2B Antibody (PHOX2B/7792R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20799JF635
This item is not returnable.
View return policy
Description
PHOX2B Monoclonal antibody specifically detects PHOX2B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PHOX2B | |
| Monoclonal | |
| Janelia Fluor 635 | |
| NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein | |
| Recombinant fragment of human PHOX2B protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Neuroscience | |
| 8929 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PHOX2B/7792R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction