missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25276-1-AP-20UL
This item is not returnable.
View return policy
Description
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.Specifications
| PHOX2B | |
| Polyclonal | |
| Unconjugated | |
| PHOX2B | |
| NBPhox, Neuroblastoma Phox, paired like homeobox 2b, PHOX2B, PHOX2B homeodomain protein, PMX2B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18935, 8929 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35690, Q99453 | |
| Phox2b | |
| PHOX2B Fusion Protein Ag13196 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction