missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHPT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121685
This item is not returnable.
View return policy
Description
PHPT1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. P97, a cell cycle regulator involved in a variety of membrane related functions, has been shown to be a substrate of this PTP. This PTP was also found to interact with, and be regulated by adaptor protein 14-3-3 beta. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PHPT1 | |
| Polyclonal | |
| Unconjugated | |
| Phpt1 | |
| 14 kDa phosphohistidine phosphatase; 14kDa phosphohistidine phosphatase; 1700008C22Rik; CGI-202; epididymis secretory sperm binding protein Li 132P; HEL-S-132P; HSPC141; hypothetical protein LOC445172; phosphohistidine phosphatase 1; PHP; PHP14; Phpt1; Protein histidine phosphatase; protein janus-A homolog; PTPH1; PTP-H1; sex-regulated protein janus-a; Unknown (protein for MGC:133682); wu:fb50e08; zgc:101086 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 29085, 296571, 75454 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9DAK9, Q9NRX4 | |
| Phpt1 | |
| Recombinant protein Phosphohistidine Phosphatase 1. The antigen corresponds to amino acid range 1-125 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction