missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHPT1 Rabbit anti-Human, Clone: 9, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 367.00 - € 552.00
Specifications
| Antigen | PHPT1 |
|---|---|
| Clone | 9 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283738
|
Novus Biologicals
NBP2-90110-50UL |
50 μg |
€ 367.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221116
|
Novus Biologicals
NBP2-90110-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHPT1 Monoclonal specifically detects PHPT1 in Human samples. It is validated for ELISA.Specifications
| PHPT1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
| Recombinant Human PHPT1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 9 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 29085 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title