Learn More
Invitrogen™ PHPT1 Recombinant Rabbit Monoclonal Antibody (009)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530747
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human PHPT1.
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. P97, a cell cycle regulator involved in a variety of membrane related functions, has been shown to be a substrate of this PTP. This PTP was also found to interact with, and be regulated by adaptor protein 14-3-3 beta. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PHPT1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9NRX4 | |
| Phpt1 | |
| Recombinant Human PHPT1 Protein (Ala2-Tyr125). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 009 | |
| Unconjugated | |
| Phpt1 | |
| 14 kDa phosphohistidine phosphatase; 14kDa phosphohistidine phosphatase; 1700008C22Rik; CGI-202; epididymis secretory sperm binding protein Li 132P; HEL-S-132P; HSPC141; hypothetical protein LOC445172; phosphohistidine phosphatase 1; PHP; PHP14; Phpt1; Protein histidine phosphatase; protein janus-A homolog; PTPH1; PTP-H1; sex-regulated protein janus-a; Unknown (protein for MGC:133682); wu:fb50e08; zgc:101086 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29085 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.