missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHRF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554103
This item is not returnable.
View return policy
Description
Immunogen sequence: SCIPSVLKPV EPSLGLLRAD IGAASLSLFG DPYELDPFDS SEELSANPLS PLSAKRRALS RSALQSHQPV ARPVSVGLSR RRLPAAVPEP DLEEEPVPDL LGSILSGQSL LMLGSSDVII HRDGSLSAKR AAPVSFQR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
Specifications
| PHRF1 | |
| Polyclonal | |
| Unconjugated | |
| PHRF1 | |
| AA673488; CTD-binding SR-like protein rA9; Kiaa1542; PHD and RING finger domain-containing protein 1; PHD and ring finger domains 1; phrf1; PPP1R125; protein phosphatase 1, regulatory subunit 125; RNF221 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P1Y6 | |
| PHRF1 | |
| Recombinant protein corresponding to Human PHRF1. Recombinant protein control fragment (Product #RP-91599). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction