missing translation for 'onlineSavingsMsg'
Learn More

Phthalamide, 97%

Product Code. p-7044881
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11372596

Brand: Thermo Scientific Alfa Aesar L08600.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-96-0 | C8H8N2O2 | 164.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-96-0
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00025478
InChI Key NAYYNDKKHOIIOD-UHFFFAOYSA-N
Synonym phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs
PubChem CID 6956
ChEBI CHEBI:38799
IUPAC Name benzene-1,2-dicarboxamide
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)N

Specifications

Melting Point 223°C (decomposition)
Quantity 10 g
Beilstein 1868220
Merck Index 14,7369
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material Phthalamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.