missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYH Antibody (03), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06164B
This item is not returnable.
View return policy
Description
PHYH Monoclonal antibody specifically detects PHYH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PHYH | |
| Monoclonal | |
| Biotin | |
| LNAP1, PAHXLN1, peroxisomal, PhyH, PHYH1, phytanoyl-CoA 2-hydroxylase, phytanoyl-CoA hydroxylase, phytanoyl-CoA hydroxylase (Refsum disease), RD | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PHYH (O14832; Ser31-Leu338). | |
| 0.1 mL | |
| Lipid and Metabolism, Vision | |
| 5264 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 03 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction