missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYHD1 Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811367
This item is not returnable.
View return policy
Description
PHYHD1 (phytanoyl-CoA dioxygenase domain containing 1) is a 291 amino acid protein belonging to the PHYH family and the PHYHD1 subfamily. Encoded by a gene that maps to human chromosome 9q34.11, PHYHD1 exists as three alternatively spliced isoforms and likely functions as an alpha-ketoglutatare-dependent dioxygenase. PHYHD1 participates in metal ion binding and oxidoreductase activity, thereby acting on single donors with incorporation of two atoms of oxygen. Related to PHYH (phytanoyl-CoA 2-hydroxylase), an Fe(II) and 2-oxoglutarate (2OG)-dependent oxygenase that catalyzes the initial alpha-oxidation step in the degradation of phytenic acid in peroxisomes, PHYHD1 similarly exhibits additional homologues in a wide range of metazoans and bacteria. PHYHD1 also may play a role in DNA methylation in early postnatal liver development and mammalian differentiation.Specifications
| PHYHD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5SRE7 | |
| PHYHD1 | |
| Full length human recombit protein of human PHYHD1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A6 | |
| Unconjugated | |
| PHYHD1 | |
| phytanoyl-CoA dioxygenase domain containing 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 254295 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction