missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYKPL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539011
This item is not returnable.
View return policy
Description
This is a nuclear gene encoding a mitochondrial enzyme that catalyzes the conversion of 5-phosphonooxy-L-lysine to ammonia, inorganic phosphate, and 2-aminoadipate semialdehyde. Mutations in this gene may cause phosphohydroxylysinuria. Alternative splicing results in multiple transcript variants. [provided by RefSeq, May 2013].
Specifications
| PHYKPL | |
| Polyclonal | |
| Unconjugated | |
| PHYKPL | |
| AGXT2L2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100169747, 72947, 85007 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IUZ5, Q8R1K4 | |
| PHYKPL | |
| A synthetic peptide derived from the internal region of human AGXT2L2 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction