Learn More
Physcion 8-beta-D-glucoside, TRC
Specifications
Specifications
| Chemical Name or Material | Physcion 8-beta-D-glucoside |
| CAS | 23451-01-6 |
| Molecular Formula | C22 H22 O10 |
| Quantity | 1 mg |
| Synonym | β-D-8-Hydroxy-3-methoxy-6-methyl-1-anthraquinonyl Glucopyranoside, Physcion 8-β-D-glucopyranoside, Physcion 8-β-D-monoglucoside, Anthraglycoside A, Physcion 8-O-β-D-glucopyranoside, 1-(β-D-glucopyranosyloxy)-8-hydroxy-3-methoxy-6-methyl-9,10-Anthracenedione |
| InChI Formula | InChI=1S/C22H22O10/c1-8-3-10-15(12(24)4-8)19(27)16-11(17(10)25)5-9(30-2)6-13(16)31-22-21(29)20(28)18(26)14(7-23)32-22/h3-6,14,18,20-24,26,28-29H,7H2,1-2H3/t14-,18-,20+,21-,22-/m1/s1 |
| SMILES | COc1cc(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)c3C(=O)c4c(O)cc(C)cc4C(=O)c3c1 |
| IUPAC Name | 1-hydroxy-6-methoxy-3-methyl-8-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-anthracene-9,10-dione |
| Molecular Weight (g/mol) | 446.4 |
| Formula Weight | 446.1213 |