missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p110 beta/PIK3CB Antibody (5A9), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | PI 3-Kinase p110 beta/PIK3CB |
|---|---|
| Clone | 5A9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30037465
|
Novus Biologicals
NBP3-26383-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032989
|
Novus Biologicals
NBP3-26383-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PI 3-Kinase p110 beta/PIK3CB Monoclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| PI 3-Kinase p110 beta/PIK3CB | |
| Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta | |
| A synthesized peptide derived from Human PI 3-Kinase p110 beta/PIK3CB [UniProt P42338] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5A9 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5291 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title