missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p110 gamma Antibody (4B5D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52531
This item is not returnable.
View return policy
Description
PI 3-Kinase p110 gamma Monoclonal specifically detects PI 3-Kinase p110 gamma in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PI 3-Kinase p110 gamma | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| 1-phosphatidylinositol 3-kinase, EC 2.7.1, EC 2.7.1.153, p110-gamma, p120-PI3K, phosphatidylinositol 3 kinase gamma, p110 gamma, phosphatidylinositol 3-kinase catalytic 110-kD gamma, phosphatidylinositol 3-kinase, catalytic, gamma polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform, phosphoinositide-3-kinase gamma catalytic subunit, phosphoinositide-3-kinase, catalytic, gamma polypeptide, PI3CG, PI3K, PI3Kgamma, PI3K-gamma, PI3-kinase subunit gamma, PIK3, PtdIns-3-kinase subunit gamma, PtdIns-3-kinase subunit p110-gamma | |
| Mouse | |
| 126.5 kDa | |
| 0.1 mg | |
| mTOR Pathway, Phospho Specific, Signal Transduction | |
| 5294 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 4B5D2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PIK3CG | |
| Purified recombinant fragment of human PIK3CG (AA: 1-100) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction