missing translation for 'onlineSavingsMsg'
Learn More

PI-3065, MedChemExpress

Product Code. 16445038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445038 10 mg 10mg
16495776 100 mg 100mg
16411277 50 mg 50mg
16407767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16445038 Supplier Medchem Express Supplier No. HY12235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PI-3065 is a potent inhibitor of PI3K p110δ, with IC50 and Ki values of 5 nM and 1.5 nM, and exhibits less potent activity against p110α, p110β, p110γ with IC50s of 910, 600, >10000 nM.

PI-3065 is a potent inhibitor of PI3K p110δ, with IC50 and Ki values of 5 nM and 1.5 nM, and exhibits less potent activity against p110α, p110β, p110γ with IC50s of 910, 600, >10000 nM.

Specifications

Chemical Name or Material PI-3065
CAS 955977-50-1
Color White
Purity Grade Notes Research
Molecular Formula C27H31FN6OS
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (49.34 mM; Need ultrasonic)
SMILES FC(C=CC1=C2C=CN1)=C2C3=NC4=C(SC(CN5CCN(CC6CC6)CC5)=C4)C(N7CCOCC7)=N3
Molecular Weight (g/mol) 506.64
Formula Weight 506.64
Percent Purity 97.26%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.