missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4K2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582133
This item is not returnable.
View return policy
Description
Immunogen sequence: LILPYISDMN FVQDLCEDLY ELFKTDKGFD KATFESQMSV MRGQILNLTQ ALRDGKSPFQ LVQIPCVIVE RSQGGSQGRI VHLSNSFTQT VNCRKPF Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 87%.
PI4K2B lipid kinase is a member of the phosphatidylinositol 4-kinase family (PI4K) that phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate which are immediate precursor of several important signaling and scaffolding molecules. PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses. PI4K2B uses phosphatidylinositol as its primary substrate and has no activity on phosphatidylinositol monophosphates. PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation. Phosphatidylinositol 4-kinases (PI4Ks) phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate (PIP), an immediate precursor of several important signaling and scaffolding molecules. PIP itself may also have direct functional and structural roles. PI4K2B is a primarily cytosolic PI4K that is recruited to membranes, where it stimulates phosphatidylinositol 4, 5-bisphosphate synthesis (Wei et al., 2002 ).
Specifications
| PI4K2B | |
| Polyclonal | |
| Unconjugated | |
| PI4K2B | |
| 2610042N09Rik; 4933409G22Rik; hypothetical protein; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 beta; Phosphatidylinositol 4-kinase type 2 beta; Phosphatidylinositol 4-kinase type 2-beta; phosphatidylinositol 4-kinase type 2-beta; Phosphatidylinositol 4-kinase type II-beta; phosphatidylinositol 4-kinase type-II beta; PI4K2B; PI4K2B protein; PI4KIIB; PI4KII-BETA; PIK42B; RCJMB04_25j21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55300 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCG2 | |
| PI4K2B | |
| Recombinant protein corresponding to Human PI4K2B. Recombinant protein control fragment (Product #RP-102316). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction