Learn More
PICALM Mouse anti-Human, Unconjugated, Clone: 1F6G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67564-1-IG-150UL
Description
The PICALM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PICALM. This antibody recognizes human, mouse, rat antigen. The PICALM antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PICALM | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F6G7 | |
| Unconjugated | |
| 150 μL | |
| 8301 | |
| IgG1 |