Learn More
VITLAB™ Piccolo 1™ Dispenser
Brand: VITLAB™ 1610501
220.77 EUR valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Additional Details : Weight : 0.14000kg
Specifications
≤±3% | |
Bottletop Dispenser | |
100 μL |