missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piccolo Antibody (6H9-B6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-49453JF646
This item is not returnable.
View return policy
Description
Piccolo Monoclonal antibody specifically detects Piccolo in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Piccolo | |
| Monoclonal | |
| Janelia Fluor 646 | |
| piccolo (presynaptic cytomatrix protein), protein piccolo | |
| Full length protein, BC001304.1 | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience | |
| 27445 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 6H9-B6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction