missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PICK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11306573
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306573 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306573 Supplier Invitrogen™ Supplier No. PA518593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKCalpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK1 with PKCalpha is highly dependent on the activation of the kinase. It appears that PICK1 directs the activated form of PKCalpha to membrane anchored GluR2. Once phosphorylated by PKCalpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK1-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PICK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Pick1
Gene Accession No. Q62083, Q9NRD5
Gene Alias MGC15204; OTTHUMP00000028509; PICK; Pick1; PRKCA-binding protein; Prkcabp; Protein interacting with C kinase 1; protein interacting with PRKCA 1; protein kinase C, alpha binding protein; protein kinase C-alpha-binding protein; protein that interacts with C kinase 1
Gene Symbols Pick1
Host Species Goat
Immunogen Synthetic peptide sequence (HYNKLQADPKQ) corresponding to the internal amino acids of PICK1 (aa 100-110).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18693, 9463
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.