Learn More
Picropodophyllin, MedChemExpress
Product Code. 16426578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426578 10 mg 10mg
16439147 5 mg 5mg
16432667 50 mg 50mg
16406786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16426578 Supplier Medchem Express Supplier No. HY15494
MedChemExpress Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.
Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.
Specifications
Chemical Name or Material Picropodophyllin
CAS 477-47-4
Color White
Purity Grade Notes Research
Molecular Formula C22H22O8
Quantity 10 mg
Synonym AXL1717 Picropodophyllin PPP
Solubility Information DMSO : 50 mg/mL (120.65 mM; Need ultrasonic)
SMILES O=C1OC[C@]2([H])[C@@H](O)C3=C(C=C4OCOC4=C3)[C@@H](C5=CC(OC)=C(OC)C(OC)=C5)[C@@]21[H]
Molecular Weight (g/mol) 414.41
Formula Weight 414.41
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only