missing translation for 'onlineSavingsMsg'
Learn More

Pictilisib, MedChemExpress

Product Code. 16401316
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401316 200 mg 200mg
16463977 50 mg 50mg
16458058 10 mg 10mg
16457836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16401316 Supplier Medchem Express Supplier No. HY50094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).

Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).

Specifications

Chemical Name or Material Pictilisib
CAS 957054-30-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27N7O3S2
Quantity 200 mg
Synonym GDC-0941
Solubility Information DMSO : 100 mg/mL (194.69 mM; ultrasonic and warming and heat to 60°C)
SMILES CS(N1CCN(CC2=CC3=C(C(N4CCOCC4)=NC(C5=CC=CC6=C5C=NN6)=N3)S2)CC1)(=O)=O
Molecular Weight (g/mol) 513.64
Formula Weight 513.64
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.