Learn More
Pictilisib, MedChemExpress
Product Code. 16463977
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463977 50 mg 50mg
16458058 10 mg 10mg
16457836 100 mg 100mg
16401316 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16463977 Supplier Medchem Express Supplier No. HY50094
MedChemExpress Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).
Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).
Specifications
Chemical Name or Material Pictilisib
CAS 957054-30-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27N7O3S2
Quantity 50 mg
Synonym GDC-0941
Solubility Information DMSO : 100 mg/mL (194.69 mM; ultrasonic and warming and heat to 60°C)
SMILES CS(N1CCN(CC2=CC3=C(C(N4CCOCC4)=NC(C5=CC=CC6=C5C=NN6)=N3)S2)CC1)(=O)=O
Molecular Weight (g/mol) 513.64
Formula Weight 513.64
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only