missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PID1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557521
This item is not returnable.
View return policy
Description
Immunogen sequence: VSPNIFAWVY REINDDLSYQ MDCHAVECES KLEAKKLAHA MMEAFRKTFH SMKSDGRIHS NSSSEEVSQE LESDDG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Increases proliferation of preadipocytes without affecting adipocytic differentiation.
Specifications
| PID1 | |
| Polyclonal | |
| Unconjugated | |
| PID1 | |
| 5033414K04Rik; Ab1-119; HMFN2073; NYGGF4; PCLI1; P-CLI1; phosphotyrosine interaction domain containing 1; phosphotyrosine interaction domain-containing protein 1; Pid1; Protein NYGGF4; PTB-containing, cubilin and LRP1-interacting protein; RGD1560766 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55022 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z2X4 | |
| PID1 | |
| Recombinant protein corresponding to Human PID1. Recombinant protein control fragment (Product #RP-96150). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction