missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIDD Polyclonal Antibody
GREENER_CHOICE

Product Code. 17720664
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17720664 200 μL 200µL
17723084 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17720664 Supplier Invitrogen™ Supplier No. PA5119255

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIDD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene contains a leucine-rich repeat and a death domain. This protein has been shown to interact with other death domain proteins, such as Fas (TNFRSF6)-associated via death domain (FADD) and MAP-kinase activating death domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIDD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PIDD1
Gene Accession No. Q9ERV7
Gene Alias 1200011D09Rik; AU042446; DKFZp434D229; leucine-rich and death domain containing; leucine-rich repeat and death domain-containing protein; leucine-rich repeats and death domain containing; Lrdd; MGC16925; p53 induced death domain protein 1; p53 protein induced, with death domain; p53-induced death domain protein 1; p53-induced death domain-containing protein 1; p53-induced protein with a death domain; Pidd; Pidd1; PIDD-C; PIDD-CC; PIDD-N
Gene Symbols PIDD1
Host Species Rabbit
Immunogen Recombinant protein Leucine Rich Repeats And Death Domain Containing Protein. The antigen corresponds to amino acid range 666-915 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57913
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.