missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pierce-1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81981H
This item is not returnable.
View return policy
Description
Pierce-1 Polyclonal antibody specifically detects Pierce-1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| Pierce-1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| C9orf116, chromosome 9 open reading frame 116, FLJ13945, hypothetical protein LOC138162, MGC29761, Pierce 1, RbEST47, RP11-426A6.4 | |
| Pierce-1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Pierce-1. The immunogen is located within the first 50 amino acids of Pierce-1. Amino Acid Squence: APKATAPPERTSDYYR | |
| 0.1 mL | |
| Apoptosis | |
| 138162 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction