Learn More
Thermo Scientific™ SIAB (succinimidyl (4-iodoacetyl)aminobenzoate)
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and iodoacetyl (haloacetyl) groups at ends of a 9-atom (10.6A) aminobenzoate spacer arm.
Brand: Thermo Scientific™ 22329
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce SIAB is a mid-length (10.6 Å) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and iodoacetyl reactive groups.
Highlights:
- Reactive groups: NHS ester and iodoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Amine-Sulfhydryl | |
SIAB | |
No | |
Upon receipt store at 4°C protected from light. | |
Ambient | |
Chemical Labeling | |
Haloacetyl, NHS Ester | |
Powder | |
DMF, DMSO | |
No |
No | |
402.14 | |
10.6 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.