Learn More
Description
These Thermo Scientific Pierce cell culture media and serum components are optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
Highlights:
Flexible–liquid media deficient in both L-lysine and L-arginine and powdered media deficient in L-leucine, L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
Compatible–DMEM, MEM, RPMI-1640, IMDM, Ham's F12 and McCoy's 5A media and dialyzed FBS are used to culture a variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3, K562 and Jurkat
Optimized, specialized media–the Expansion DMEM and Low-Osmolarity DMEM are specifically designed and qualified for mouse embryonic stem cell propagation and SILAC protein expression profiling with mass spectrometry
High quality–Stem Cell Screened Dialyzed FBS is tested to ensure that it does not promote stem cell differentiation
Specifications
Specifications
| Description | DMEM Media for SILAC, 6 x 500mL, Cells Starting Material, Used With Mass Spectrometer, For Use With Stable Isotope Labeling With Amino Acids in Cell Culture (SILAC) To Analyze Protein Expression by Mass Spectrometry (MS) |
| Formulation | DMEM media minus L-Lysine and L-Arginine |
| For Use With (Equipment) | Mass spectrometry |
| Product Type | DMEM |
| Sufficient For | 1 to 3 SILAC experiments |
| Content And Storage | 2°C to 8°C |
| Quantity | 6 x 500 mL |
| For Use With (Application) | Protein biology, protein quantitation using mass spectrometry, protein mass spectrometry analysis, SILAC experiments (stable isotope labeling with amino acids in cell culture) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.