Learn More
New Pig Lite-Dri™ Loose Absorbent
Product Code. 12973625
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
22 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
12973625 22 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12973625 Supplier New Pig Supplier No. PLP201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Use silica-free material to reduce labor and disposal costs
  • Silica-free composition is non-abrasive and does not pose the health hazards associated with breathing in clay dust
  • Non-abrasive composition won't harm finished floors or damage expensive machinery
  • Light-weight absorbent is easier than clay to carry and use without injury
  • Absorbs and retains oils, coolants, solvents and water
Specifications
Capacity (English) 8 gal.
Capacity (Metric) 30.28 L
Material Cellulose
Type Loose Absorbent
Format Powder
For Use With (Application) For most common industrial liquids, is easier to lift and lighter to carry than clay
Packaging Quantity 22 lb.
Length (English) 29 in.
Length (Metric) 73.66 cm
Width (English) 6 in.
Width (Metric) 15.24 cm
Weight (English) 23 lb.
Weight (Metric) 10 kg