missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIG3 Antibody (OTI3B11), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73400B
This item is not returnable.
View return policy
Description
PIG3 Monoclonal specifically detects PIG3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PIG3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TP53I3 | |
| Full length recombinant protein of human PIG3 (NP_671713) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3B11 | |
| Western Blot, Immunohistochemistry | |
| p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 9540.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction