missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553409
This item is not returnable.
View return policy
Description
Immunogen sequence: VEYDGTTSFF VSDYTEVDNN VTRHLDKVLK RGDWDILILH YLGLDHIGHI SGPNSPLIGQ KLSEMDSVLM KIHTSLQSKE RETPLPNLLV LCGDHGMSET GSHGASSTEE VNTPLILISS AFERKPGDIR HPKHVQQTDV A Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Specifications
| PIGG | |
| Polyclonal | |
| Unconjugated | |
| PIGG | |
| GPI ethanolamine phosphate transferase 2; GPI7; GPI7 homolog; hGPI7; LAS21; MRT53; phosphatidylinositol glycan anchor biosynthesis class G; phosphatidylinositol glycan anchor biosynthesis, class G; phosphatidylinositol-glycan biosynthesis class G protein; PIGG; PIG-G; PRO4405; RLGS1930; UNQ1930/PRO4405 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5H8A4 | |
| PIGG | |
| Recombinant protein corresponding to Human PIGG. Recombinant protein control fragment (Product #RP-91112). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction