missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556802
This item is not returnable.
View return policy
Description
Immunogen sequence: KIDQETLLII DSLGIQMTSS YASGKESTTF IEMGKVKDIV INEAIYMQKV IYYLCILLKD PVEPHGISQV VPVFQSAKPR LDCLIEVYRS CQEILAHQKA TSTS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. The protein encoded by this gene is a subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. [provided by RefSeq, Jul 2008].
Specifications
| PIGH | |
| Polyclonal | |
| Unconjugated | |
| PIGH | |
| 2210416H01Rik; A930028P05Rik; GPI-H; Mrp64; phosphatidylinositol glycan anchor biosynthesis class H; phosphatidylinositol glycan anchor biosynthesis, class H; phosphatidylinositol glycan, class H; phosphatidylinositol N-acetylglucosaminyltransferase subunit H; Phosphatidylinositol-glycan biosynthesis class H protein; phosphatidylinositol-glycan biosynthesis, class H protein; Pigh; PIG-H | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14442 | |
| PIGH | |
| Recombinant protein corresponding to Human PIGH. Recombinant protein control fragment (Product #RP-89081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction