missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIGK Polyclonal Antibody
GREENER_CHOICE

Product Code. 16521522
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16521522 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16521522 Supplier Invitrogen™ Supplier No. PA598697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIGK Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the cysteine protease family C13 that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is a member of the multisubunit enzyme, GPI transamidase and is thought to be its enzymatic component. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PIGK
Gene Accession No. Q92643
Gene Alias 3000001O05Rik; GPI transamidase; GPI transamidase subunit; GPI8; GPI8 homolog; GPI-anchor transamidase; hGPI8; phosphatidylinositol glycan anchor biosynthesis class K; phosphatidylinositol glycan anchor biosynthesis, class K; phosphatidylinositol glycan, class K; phosphatidylinositol-glycan biosynthesis class K protein; Pigk; PIG-K
Gene Symbols PIGK
Host Species Rabbit
Immunogen Recombinant Human GPI-anchor transamidase protein (91-293AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10026
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.