missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pIgR Antibody (007), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89286CL1
This item is not returnable.
View return policy
Description
pIgR Monoclonal antibody specifically detects pIgR in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| pIgR | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human pIgR (Accession#: NP_002635.2; Met1-Arg 638,365 Ser/Gly). | |
| 0.1 mL | |
| Immunology | |
| 5284 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 007 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction