missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGU Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559685
This item is not returnable.
View return policy
Description
Immunogen sequence: KLLLELDQYA PDVAELIRTP MEMRYIPLKV ALFYLLNPYT ILSCVAKSTC AINN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Component of the GPI transamidase complex. May be involved in the recognition of either the GPI attachment signal or the lipid portion of GPI.
Specifications
| PIGU | |
| Polyclonal | |
| Unconjugated | |
| PIGU | |
| 5430426F17Rik; BC028278; CDC91 (cell division cycle 91, S. cerevisiae, homolog)-like 1; CDC91 cell division cycle 91-like 1; CDC91L1; cell division cycle 91-like 1; cell division cycle 91-like 1 protein; cell division cycle protein 91-like 1; GAB1; GPI transamidase component PIG-U; GPI transamidase subunit; liver regeneration-related protein LRRGT00059; phosphatidylinositol glycan anchor biosynthesis class U; phosphatidylinositol glycan anchor biosynthesis class U protein; phosphatidylinositol glycan anchor biosynthesis, class U; phosphatidylinositol-glycan biosynthesis, class U protein; Pigu; Protein CDC91-like 1; PSEC0205; sakura; UNQ3055/PRO9875 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H490 | |
| PIGU | |
| Recombinant protein corresponding to Human PIGU. Recombinant protein control fragment (Product #RP-99351). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction