missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIGV Polyclonal Antibody
Description
Peptide sequence: FVDQLVEGLL GGLSHWDAEH FLFIAEHGYL YEHNFAFFPG FPLALLVGTE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Glycosylphosphatidylinositol (GPI) is a complex glycolipid that anchors many proteins to the cell surface. The biosynthetic pathway of GPI is mediated by sequential addition of sugars and other components to phosphatidylinositol. PIGV adds the second mannose to the GPI core.
Specifications
Specifications
| Antigen | PIGV |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PIGV |
| Gene Accession No. | Q9NUD9 |
| Gene Alias | B330013B03; D430024F16Rik; dol-P-Man dependent GPI mannosyltransferase; FLJ20477; GPI mannosyltransferase 2; GPI mannosyltransferase II; GPI-MT-II; HPMRS1; LOW QUALITY PROTEIN: GPI mannosyltransferase 2; phosphatidylinositol glycan anchor biosynthesis class V; phosphatidylinositol glycan anchor biosynthesis, class V; phosphatidylinositol glycan class V; phosphatidylinositol glycan, class V; phosphatidylinositol-glycan biosynthesis class V protein; Pigv; PIG-V; RGD1309526; Ybr004c homolog |
| Gene Symbols | PIGV |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?