missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGZ Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93538-0.02ml
This item is not returnable.
View return policy
Description
PIGZ Polyclonal antibody specifically detects PIGZ in Human samples. It is validated for Western Blot
Specifications
| PIGZ | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| class Z, EC 2.4.1, EC 2.4.1.-, FLJ12768, GPI mannosyltransferase 4, GPI mannosyltransferase IV, GPI-MT-IV, hSMP3, phosphatidylinositol glycan anchor biosynthesis, class Z, Phosphatidylinositol-glycan biosynthesis class Z protein, PIG-Z, SMP3 mannosyltransferase, SMP3SMP3 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 435-579 of human PIGZ (NP_079439.2). LVPGLEYLEQVVHAPVLPSTPTHYTLLFTHTYMPPRHLLHLPGLGAPVEVVDMGGTEDWALCQTLKSFTRQPACQVAGGPWLCRLFVVTPGTTRRAVEKCSFPFKNETLLFPHLTLEDPPALSSLLSGAWRDHLSLHIVELGEET | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 80235 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction