missing translation for 'onlineSavingsMsg'
Learn More

PIH1D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7208994 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16818994 20 μL 20µL
16808994 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16818994 Supplier Proteintech Supplier No. 194271AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Involved in the assembly of C/D box small nucleolar ribonucleoprotein (snoRNP) particles. Recruits the SWI/SNF complex to the core promoter of rRNA genes and enhances pre-rRNA transcription. Mediates interaction of TELO2 with the R2TP complex which is necessary for the stability of MTOR and SMG1. Positively regulates the assembly and activity of the mTORC1 complex.

Specifications

Antigen PIH1D1
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PIH1D1
Gene Accession No. Q4V7F5, Q9CQJ2, Q9NWS0
Gene Alias NOP17, Nucleolar protein 17 homolog, PIH1 domain containing 1, PIH1D1
Gene Symbols Pih1d1
Host Species Rabbit
Immunogen PIH1D1 Fusion Protein Ag13759
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292898, 55011, 68845
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.