missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIH1D2 Antibody (OTI4A10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73403
This item is not returnable.
View return policy
Description
PIH1D2 Monoclonal specifically detects PIH1D2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| PIH1D2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
| PIH1 domain containing 2, PIH1 domain-containing protein 2 | |
| Mouse | |
| 35.8 kDa | |
| 100 μg | |
| 120379 | |
| Human | |
| Lyophilized |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| OTI4A10 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PIH1D2 | |
| Human recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2(NP_620144) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction