missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIH1D2 Antibody (OTI4A10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73403H
This item is not returnable.
View return policy
Description
PIH1D2 Monoclonal specifically detects PIH1D2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PIH1D2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PIH1D2 | |
| Recombinant protein fragment corresponding to amino acids 1-315 of human PIH1D2 (NP_620144) produced in E.coli. | |
| 0.1 mL | |
| 120379.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4A10 | |
| Western Blot, Immunohistochemistry | |
| PIH1 domain containing 2, PIH1 domain-containing protein 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction