missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3AP1 Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501716
This item is not returnable.
View return policy
Description
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.Specifications
| PIK3AP1 | |
| Monoclonal | |
| 0.63 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZUJ8 | |
| PIK3AP1 | |
| Full length human recombit protein of human PIK3AP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| PIK3AP1 | |
| BCAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 118788 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction