Learn More
PIK3AP1 Monoclonal Antibody (OTI7A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501812
This item is not returnable.
View return policy
Description
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.Specifications
| PIK3AP1 | |
| Monoclonal | |
| 0.98 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZUJ8 | |
| PIK3AP1 | |
| Full length human recombit protein of human PIK3AP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7A11 | |
| Unconjugated | |
| PIK3AP1 | |
| BCAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 118788 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |