missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3C2A Antibody (OTI3H2), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73405V
This item is not returnable.
View return policy
Description
PIK3C2A Monoclonal specifically detects PIK3C2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PIK3C2A | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PIK3C2A | |
| Recombinant protein fragment corresponding to amino acids 230-560 of human PIK3C2A (NP_002636) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3H2 | |
| Western Blot, Immunohistochemistry | |
| C2-containing phosphatidylinositol kinase, CPK, DKFZp686L193, EC 2.7.1, EC 2.7.1.154, MGC142218, phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha, Phosphoinositide 3-kinase-C2-alpha, phosphoinositide-3-kinase, class 2, alpha polypeptide, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2alpha, PI3K-C2-alpha, PtdIns-3-kinase C2 subunit alpha | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 5286.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction