missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIK3C2A Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801708S
This item is not returnable.
View return policy
Description
PIK3C2A Monoclonal antibody specifically detects PIK3C2A in Human samples. It is validated for Western BlotSpecifications
| PIK3C2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00987 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4193 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI2B5 | |
| Unconjugated | |
| MDM2 | |
| C2-containing phosphatidylinositol kinase; CPK; Cpk-m; DKFZp686L193; MGC142218; p170; phosphatidylinositol 3-kinase, C2 domain containing, alpha polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit alpha; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha; phosphoinositide 3-kinase-C2-alpha; phosphoinositide-3-kinase, class 2, alpha polypeptide; PI3KC2; PI3-K-C2(ALPHA); pi3kc2a; PI3-K-C2A; PI3K-C2alpha; PI3K-C2-alpha; Pik3c2a; ptdIns-3-kinase C2 subunit alpha | |
| Human recombinant protein fragment corresponding to amino acids 119-438 of human MDM2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction