missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT2 Mouse anti-Human, Clone: OTI5A12, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808626S
This item is not returnable.
View return policy
Description
CNOT2 Monoclonal antibody specifically detects CNOT2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CNOT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZN8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4848 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A12 | |
| Unconjugated | |
| Cnot2 | |
| 2600016M12Rik; 2810470K03Rik; AA537049; AA959607; AW557563; C79650; CCR4-associated factor 2; CCR4-NOT transcription complex subunit 2; CCR4-NOT transcription complex, subunit 2; CDC36; CNOT2; HSPC131; MSTP046; negative regulator of transcription 2; NOT2; NOT2H | |
| Full length human recombinant protein of human CNOT2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction